Läs mer
2-(4-Chlorobutoxy)tetrahydropyran, TRC
Produktkod. 30362580
Change view
Kvantitet:
10 g
Produktkod. Kvantitet
30362580 10 g
1 options
Produktkod. 30362580 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC36444510G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(4-Chlorobutoxy)tetrahydropyran
CAS 41302-05-0
Molekylformel C9H17ClO2
Kvantitet 10 g
Synonym 2-(4-Chlorobutoxy)tetrahydro-pyran, 1-Chloro-4-(tetrahydropyranyloxy)butane, 2-(4-Chlorobutoxy)tetrahydro-2H-pyran, 2-(4-Chlorobutyloxy)tetrahydropyran, 4-(Tetrahydro-2-pyranyloxy)-1-chlorobutane, 4-Chloro-1-(tetrahydropyranyloxy)butane, 4-Chloro-1-butanol Tetrahydropyranyl Ether, 4-Chlorobutanol Tetrahydropyranyl Ether, 4-Chlorobutyl Tetrahydropyranyl Ether
InChI formel InChI=1S/C9H17ClO2/c10-6-2-4-8-12-9-5-1-3-7-11-9/h9H,1-8H2
LEDER ClCCCCOC1CCCCO1
IUPAC-namn 2-(4-chlorobutoxy)oxane
Molekylvikt (g/mol) 192.68
Formel vikt 192.0917