Läs mer
2-(4-Chlorophenyl)-3-methylbutanoic Acid, TRC
Produktkod. 30138380
Change view
Kvantitet:
1 g
2.5 g
5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30138380 2.5 g 2.5g
30137090 5 g -
30012497 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138380 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3754282.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2012-74-0
Molekylformel C11 H13 Cl O2
Molekylvikt (g/mol) 212.67
Synonym Butyric acid, 2-(p-chlorophenyl)-3-methyl- (7CI,8CI), 4-Chloro-α-(1-methylethyl)benzeneacetic acid, (RS)-2-(4-Chlorophenyl)-3-methylbutyric acid, (RS)-Fenvaleric acid, (±)-2-(4-Chlorophenyl)-3-methylbutanoic acid, (±)-2-(4-Chlorophenyl)-3-methylbutyric acid, (±)-2-(4-Chlorophenyl)isovaleric acid, (±)-2-(p-Chlorophenyl)-3-methylbutyric acid, (±)-Fenvaleric acid, (±)-α-Isopropyl-4-chlorophenylacetic acid, 2-(4-Chlorophenyl)-3-methylbutanoic acid, 2-(4-Chlorophenyl)-3-methylbutyric acid, 2-(4-Chlorophenyl)isovaleric acid, 2-(p-Chlorophenyl)-3-methylbutyric acid, 4-Chloro-α-isopropylbenzeneacetic acid, 4-Chloro-α-isopropylphenylacetic acid, Racemic fenvaleric acid, dl-2-(4-Chlorophenyl)-3-methylbutyric acid
IUPAC-namn 2-(4-chlorophenyl)-3-methylbutanoic acid
LEDER CC(C)C(C(=O)O)c1ccc(Cl)cc1
Specifikationer
Kemiskt namn eller material Fenvalerate free acid metabolite
Kvantitet 2.5 g
InChI formel InChI=1S/C11H13ClO2/c1-7(2)10(11(13)14)8-3-5-9(12)6-4-8/h3-7,10H,1-2H3,(H,13,14)
Formel vikt 212.0604