Läs mer
2,4-D Butyl Ester, TRC
Produktkod. 30378096
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30378096 250 mg
1 options
Produktkod. 30378096 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD826515250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,4-D-1-butyl ester
CAS 94-80-4
Molekylformel C12 H14 Cl2 O3
Kvantitet 250 mg
Synonym Acetic acid, (2,4-dichlorophenoxy)-, butyl ester (6CI,8CI,9CI), 2,4-D Butyl ester, 2,4-D Butylate, 2,4-DBE, 2,4-Dichlorophenoxyacetic acid butyl ester, 2,4-Dichlorophenoxyacetic acid n-butyl ester, Butapon, Butyl (2,4-dichlorophenoxy)acetate, Butyl 2,4-D, Butyl dichlorophenoxyacetate, Esso Herbicide 10, Esteron 400BR, Fernesta, Hi-Ester 2,4-D, Lironox, NSC 409767
InChI formel InChI=1S/C12H14Cl2O3/c1-2-3-6-16-12(15)8-17-11-5-4-9(13)7-10(11)14/h4-5,7H,2-3,6,8H2,1H3
LEDER CCCCOC(=O)COc1ccc(Cl)cc1Cl
IUPAC-namn butyl 2-(2,4-dichlorophenoxy)acetate
Molekylvikt (g/mol) 277.14
Formel vikt 276.032