Läs mer
2,4'-DDE, TRC
Produktkod. 30009049
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009049 500 mg
30008354 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009049 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD195200500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3424-82-6
Molekylformel C14 H8 Cl4
Molekylvikt (g/mol) 318.03
Synonym Ethylene, 1,1-dichloro-2-(o-chlorophenyl)-2-(p-chlorophenyl)- (6CI,8CI), 1-Chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene, 1,1-Dichloro-2-(o-chlorophenyl)-2-(p-chlorophenyl)ethylene, 2,4'-DDE, 2,4'-Dichlorodiphenyldichloroethylene, 2-(2-Chlorophenyl)-2-(4-chlorophenyl)-1,1-dichloroethylene, NSC 59908, o,p'-DDE, o,p'-Dichlorodiphenyldichloroethylene
IUPAC-namn 1-chloro-2-[2,2-dichloro-1-(4-chlorophenyl)ethenyl]benzene
LEDER ClC(=C(c1ccc(Cl)cc1)c2ccccc2Cl)Cl
Specifikationer
Kemiskt namn eller material 2,4'-DDE
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C14H8Cl4/c15-10-7-5-9(6-8-10)13(14(17)18)11-3-1-2-4-12(11)16/h1-8H
Formel vikt 315.938
Procent renhet >95%