Läs mer
2,4-Diamino-6-(hydroxymethyl)pteridine Hydrochloride, TRC
Produktkod. 30350042
Change view
Kvantitet:
1 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30350042 50 g
30367550 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30350042 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41639550G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 73978-41-3
Molekylformel C7 H8 N6 O . Cl H
Molekylvikt (g/mol) 228.64
Synonym (2,4-Diaminopteridin-6-yl)methanol Hydrochloride, Methotrexate Imp. A (EP) as Hydrochloride
IUPAC-namn (2,4-diaminopteridin-6-yl)methanol;hydrochloride
LEDER Cl.Nc1nc(N)c2nc(CO)cnc2n1
Specifikationer
Kemiskt namn eller material (2,4-Diaminopteridin-6-yl)methanol Hydrochloride
Kvantitet 50 g
InChI formel InChI=1S/C7H8N6O.ClH/c8-5-4-6(13-7(9)12-5)10-1-3(2-14)11-4;/h1,14H,2H2,(H4,8,9,10,12,13);1H
Formel vikt 228.0526