Läs mer
(2,4-Diaminopyrimidin-5-yl)-(3,4,5-trimethoxyphenyl)methanone, Mikromol™
Produktkod. 30050981
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30050981 25 mg
17972816 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30050981 Leverantör Mikromol Leverantörsnummer MM0093.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2,4-Diaminopyrimidin-5-yl)-(3,4,5-trimethoxyphenyl)methanone
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2,4-diaminopyrimidin-5-yl)(3,4,5-trimethoxyphenyl)methanone
CAS 30806-86-1
Analyt- eller komponentnamn (2,4-diaminopyrimidin-5-yl)(3,4,5-trimethoxyphenyl)methanone
Frakt skick Room Temperature
Molekylformel C14 H16 N4 O4
Certifikat och efterlevnad ISO 17025
Synonym Trimethoprim Imp. B (EP), Trimethoprim USP Related Compound B, Trimethoprim USP RC B, (2,4-Diaminopyrimidin-5-yl)-(3,4,5-trimethoxyphenyl)methanone, 115U67, GW 382217X, BP Trimethoprim Impurity B, Ph Eur Trimethoprim Impurity B, Trimethoprim Related Compound B, Trimethoprim Impurity B
InChI formel InChI=1S/C14H16N4O4/c1-20-9-4-7(5-10(21-2)12(9)22-3)11(19)8-6-17-14(16)18-13(8)15/h4-6H,1-3H3,(H4,15,16,17,18)
LEDER COc1cc(cc(OC)c1OC)C(=O)c2cnc(N)nc2N
IUPAC-namn (2,4-diaminopyrimidin-5-yl)-(3,4,5-trimethoxyphenyl)methanone
Molekylvikt (g/mol) 304.3
Kvantitet 25 mg
Formel vikt 304.1172 g/mol