missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-6-chloroaniline, 97%, Thermo Scientific™

Catalog Number p-7024404
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
874-18-0
C6H4Br2ClN
285.363
MFCD00031419
MFROLNRQDCAZCD-UHFFFAOYSA-N
2-chloro-4,6-dibromoaniline, 6-chloro-2,4-dibromoaniline, 4,6-dibromo-2-chlorophenylamine, pubchem3631, acmc-209qkg, 4,6-dibromo-2-chloroaniline, benzenamine,2,4-dibromo-6-chloro, labotest-bb lt01147812, 2,4-dibromo-6-chloroaniline, 2,4-bis bromanyl-6-chloranyl-aniline
4295982
2,4-dibromo-6-chloroaniline
C1=C(C=C(C(=C1Br)N)Cl)Br
This item is not returnable. View return policy
874-18-0
C6H4Br2ClN
285.363
MFCD00031419
MFROLNRQDCAZCD-UHFFFAOYSA-N
2-chloro-4,6-dibromoaniline, 6-chloro-2,4-dibromoaniline, 4,6-dibromo-2-chlorophenylamine, pubchem3631, acmc-209qkg, 4,6-dibromo-2-chloroaniline, benzenamine,2,4-dibromo-6-chloro, labotest-bb lt01147812, 2,4-dibromo-6-chloroaniline, 2,4-bis bromanyl-6-chloranyl-aniline
4295982
2,4-dibromo-6-chloroaniline
C1=C(C=C(C(=C1Br)N)Cl)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 99°C to 101°C
Quantity 5 g
Beilstein 2718195
Formula Weight 285.38
Percent Purity 97%
Chemical Name or Material 2,4-Dibromo-6-chloroaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'