missing translation for 'onlineSavingsMsg'
Learn More

2,4'-Dibromoacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-3778148
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
99-73-0
C8H6Br2O
277.94
MFCD00000200
FKJSFKCZZIXQIP-UHFFFAOYSA-N
2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4-bromophenacyl bromide, p-bromophenacyl bromide, ethanone, 2-bromo-1-4-bromophenyl, 4'-bromophenacyl bromide, p-bromophenacyl-8, alpha,p-dibromoacetophenone, alpha,4-dibromoacetophenone, 4-bromo bromoacetyl benzene
7454
2-bromo-1-(4-bromophenyl)ethanone
C1=CC(=CC=C1C(=O)CBr)Br
This item is not returnable. View return policy
99-73-0
C8H6Br2O
277.94
MFCD00000200
FKJSFKCZZIXQIP-UHFFFAOYSA-N
2,4'-dibromoacetophenone, 2-bromo-1-4-bromophenyl ethanone, 4-bromophenacyl bromide, p-bromophenacyl bromide, ethanone, 2-bromo-1-4-bromophenyl, 4'-bromophenacyl bromide, p-bromophenacyl-8, alpha,p-dibromoacetophenone, alpha,4-dibromoacetophenone, 4-bromo bromoacetyl benzene
7454
2-bromo-1-(4-bromophenyl)ethanone
C1=CC(=CC=C1C(=O)CBr)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 107°C to 111°C
Color Yellow to Beige
Boiling Point 1415°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4COCH2Br
Quantity 10 g
Beilstein 07, 285
Fieser 01,77; 03,34
Merck Index 15, 1438
Solubility Information Solubility in water: insoluble. Other solubilities: very soluble in warm alcohol,soluble in ethyl alcohol,ethyl ether,chloroform
Formula Weight 277.94
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 2, 4'-Dibromoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2,4'-Dibromoacetophenone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'