missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-1-(2-iodophenoxy)benzene, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95%+ |
Packaging | Amber Glass Bottle |
Linear Formula | Cl2C6H3OC6H4I |
Quantity | 1 g |
Formula Weight | 364.99 |
Percent Purity | 95% |
Chemical Name or Material | 2, 4-Dichloro-1-(2-iodophenoxy)benzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'