missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-3-cyanopyridine, 95%, Thermo Scientific Chemicals

Catalog Number p-7058173
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
180995-12-4
C6H2Cl2N2
172.996
MFCD09750302
OQYZEFBWEXEOPL-UHFFFAOYSA-N
21997828
2,4-dichloropyridine-3-carbonitrile
C1=CN=C(C(=C1Cl)C#N)Cl
This item is not returnable. View return policy
180995-12-4
C6H2Cl2N2
172.996
MFCD09750302
OQYZEFBWEXEOPL-UHFFFAOYSA-N
21997828
2,4-dichloropyridine-3-carbonitrile
C1=CN=C(C(=C1Cl)C#N)Cl

2,4-Dichloro-3-cyanopyridine, is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloro-3-cyanopyridine, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3439
Synonym 2,4-dichloro-3-cyanopyridine, 2,4-dichloronicotinonitrile, 2,4-dichloro-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 2,4-dichloro, 3-cyano-2,4-dichloropyridine, pubchem5457, bys83, 3-cyano-2,4dichloropyridine, ksc493a5f, 3-pyridinecarbonitrile,2,4-dichloro
Solubility Information Slightly soluble in water.
Formula Weight 173
Percent Purity 95%
Chemical Name or Material 2,4-Dichloro-3-cyanopyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'