Learn More
2,4-Dichloro-6,7-dimethoxyquinazoline, 97%, Thermo Scientific Chemicals
Catalog Number p-105129
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
27631-29-4
MFCD00051733
DGHKCBSVAZXEPP-UHFFFAOYSA-N
520327
2,4-dichloro-6,7-dimethoxyquinazoline
COC1=C(C=C2C(=C1)C(=NC(=N2)Cl)Cl)OC
27631-29-4
MFCD00051733
DGHKCBSVAZXEPP-UHFFFAOYSA-N
520327
2,4-dichloro-6,7-dimethoxyquinazoline
COC1=C(C=C2C(=C1)C(=NC(=N2)Cl)Cl)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 172°C to 177°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C10H8Cl2N2O2
Quantity 5 g
Synonym 2,4-dichloro-6,7-dimethoxy-quinazoline, 2,4-dichloro-6,7-dimethoxy quinazoline, quinazoline, 2,4-dichloro-6,7-dimethoxy, 2-chloro-6,7-dimethoxyquinazolin-4-yl chloride, quinazoline,2,4-dichloro-6,7-dimethoxy, doxazosin impurity e, doxazosin mesilate impurity e ep, pubchem9430, usp doxazosin related compound e usp, acmc-209gyd
Molecular Weight (g/mol) 259.09
Formula Weight 259.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Dichloro-6, 7-dimethoxyquinazoline, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,4-Dichloro-6,7-dimethoxyquinazoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.