missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-6-(trifluoromethyl)pyrimidine, 95%, Thermo Scientific™

Catalog Number p-4812676
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
16097-64-6
C5HCl2F3N2
216.98
ZTNFYAJHLPMNSN-UHFFFAOYSA-N
2,4-dichloro-6-trifluoromethyl pyrimidine, pyrimidine, 2,4-dichloro-6-trifluoromethyl, 2,4-dichloro-6-trifluoromethyl-pyrimidine, pubchem13247, ksc496a7p, 2,6-dichloro-4-trifluoromethyl pyrimidine
334203
2,4-dichloro-6-(trifluoromethyl)pyrimidine
C1=C(N=C(N=C1Cl)Cl)C(F)(F)F
This item is not returnable. View return policy
16097-64-6
C5HCl2F3N2
216.98
ZTNFYAJHLPMNSN-UHFFFAOYSA-N
2,4-dichloro-6-trifluoromethyl pyrimidine, pyrimidine, 2,4-dichloro-6-trifluoromethyl, 2,4-dichloro-6-trifluoromethyl-pyrimidine, pubchem13247, ksc496a7p, 2,6-dichloro-4-trifluoromethyl pyrimidine
334203
2,4-dichloro-6-(trifluoromethyl)pyrimidine
C1=C(N=C(N=C1Cl)Cl)C(F)(F)F
Boiling Point 207°C
Flash Point 78°C
Assay Percent Range 94% min. (GC)
Quantity 250 mg
Formula Weight 216.98
Percent Purity 95%
Chemical Name or Material 2, 4-Dichloro-6-(trifluoromethyl)pyrimidine
compliance-icons
Product Identifier
  • 2,4-Dichloro-6-(trifluoromethyl)pyrimidine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'