Läs mer
2,4'-Dichlorodiphenyltrichloroethane, TRC
Produktkod. 30374862
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30374862 10 mg
1 options
Produktkod. 30374862 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43420510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,4'-DDT
CAS 789-02-6
Molekylformel C14 H9 Cl5
Kvantitet 10 mg
Synonym Ethane, 1,1,1-trichloro-2-(o-chlorophenyl)-2-(p-chlorophenyl)- (6CI,8CI), 1-Chloro-2-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene, (±)-1-(o-Chlorophenyl)-1-(p-chlorophenyl)-2,2,2-trichloroethane, (±)-o,p'-DDT, (±)-o,p'-DDT, 1,1,1-Trichloro-2-(o-chlorophenyl)-2-(p-chlorophenyl)ethane, 1-(2-Chlorophenyl)-1-(4-chlorophenyl)-2,2,2-trichloroethane, 1-(o-Chlorophenyl)-1-(p-chlorophenyl)-2,2,2-trichloroethane, 2,4'-DDT, 2,4'-Dichlorodiphenyltrichloroethane, 2-(2-Chlorophenyl)-2-(4-chlorophenyl)-1,1,1-trichloroethane, 2-o-Chlorophenyl-2-p-chlorophenyl-1,1,1-trichloroethane, NSC 33446, NSC 57644, o,p'-Chlorophenothane, o,p'-DDT, o,p'-Dichlorodiphenyltrichloroethane
InChI formel InChI=1S/C14H9Cl5/c15-10-7-5-9(6-8-10)13(14(17,18)19)11-3-1-2-4-12(11)16/h1-8,13H
LEDER Clc1ccc(cc1)C(c2ccccc2Cl)C(Cl)(Cl)Cl
IUPAC-namn 1-chloro-2-[2,2,2-trichloro-1-(4-chlorophenyl)ethyl]benzene
Molekylvikt (g/mol) 354.49
Formel vikt 351.9147