Läs mer
2,4-Dichlorophenol-13C6, TRC
Produktkod. 30137833
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137833 10 mg
30135602 25 mg
30005153 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137833 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43568610MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1202864-83-2
Molekylformel 13C6 H4 Cl2 O
Molekylvikt (g/mol) 168.96
Synonym 2,4-Dichlorophenol 13C6, 2,4-Dichlorophenol-1,2,3,4,5,6-13C6
IUPAC-namn 2,4-dichloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-trien-1-ol
LEDER O[13c]1[13cH][13cH][13c](Cl)[13cH][13c]1Cl
Specifikationer
Kemiskt namn eller material 2,4-Dichlorophenol13C6
Kvantitet 10 mg
Anteckningar om renhetsgrad GC
InChI formel InChI=1S/C6H4Cl2O/c7-4-1-2-6(9)5(8)3-4/h1-3,9H/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 167.984
Procent renhet >95%