Läs mer
2,4-Dichlorophenyl Acetic Acid Methyl Ester, SPEX CertiPrep™
Produktkod. 12919184 Handla allt SPEX Certiprep Produkter
Change view
Kvantitet:
1 mL
Förpackningsstorlek:
1ml
Produktkod. Kvantitet unitSize
12919184 1 mL 1ml
1 options
Produktkod. 12919184 Leverantör SPEX Certiprep Leverantörsnummer S1421
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Innehåller
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
AnalyteCAS No.Concentration
2,4-Dichlorophenyl acetic acid methyl ester (DCAA Methyl Ester)55954-23-91000μg/mL
Certifikat
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Uppfyllande
GHS/OSHA Complaint
Specifikationer
Kemiskt namn eller material 2,4-Dichlorophenyl acetic acid methyl ester
Förpackning 2mL Amber Prescored Glass Ampule
CAS 55954-23-9, 67-56-1
CAS Min % 0.1, 99.9
Anteckningar om koncentration eller sammansättning Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylformel C9H8Cl2O2
För användning med (utrustning) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI-nyckel SRCVNBFTEARRJY-UHFFFAOYSA-N
LEDER COC(=O)CC1=C(C=C(C=C1)Cl)Cl
IUPAC-namn methyl 2-(2,4-dichlorophenyl)acetate
Lösningsmedel eller matris Methanol
Molekylvikt (g/mol) 219.061
PubChem CID 2734106
Kvantitet 1 mL
Visa mer Visa mindre
Product intended for laboratory use only