Learn More
2,4-Dichlorophenylacetic acid, 98%, BLD Pharm™
Product Code. 31068455
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31068455 1 kg
31067333 10 g
31067071 25 g
31058269 100 g
31056120 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 31068455 Supplier BLD Pharm Supplier No. BD109441000g
 Restricted Item
Beräknad leverans: 27-11-2026
Specifications
CAS 19719-28-9
Molecular Formula C8H6Cl2O2
MDL Number MFCD00004318
Quantity 1 kg
SMILES C1=C(C(=CC=C1Cl)CC(O)=O)Cl
Molecular Weight (g/mol) 205.04
Percent Purity 98%
Chemical Name or Material 2,4-Dichlorophenylacetic acid