Läs mer
2,4-Dichloroquinoline, TRC
Produktkod. 30379115
Change view
Kvantitet:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30379115 25 g
30370813 1 g
30369564 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30379115 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD43622525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 703-61-7
Molekylformel C9H5Cl2N
Molekylvikt (g/mol) 198.05
Synonym 2,4-Dichlorochinoline, 2,4-Dichloroquinoline
LEDER Clc1cc(Cl)c2ccccc2n1
Specifikationer
Kemiskt namn eller material 2,4-Dichloroquinoline
Kvantitet 25 g
InChI formel InChI=1S/C9H5Cl2N/c10-7-5-9(11)12-8-4-2-1-3-6(7)8/h1-5H
Formel vikt 196.9799