Läs mer
2,4-Difluoro-5-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}aniline, TRC
Produktkod. 30374610
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30374610 500 mg
30370944 50 mg
30370960 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30374610 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD459323500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 941166-01-4
Molekylformel C13H14N4F2
Molekylvikt (g/mol) 264.27
LEDER Nc1cc(c(F)cc1F)c2nnc3CCCCCn23
Specifikationer
Kemiskt namn eller material 2,4-Difluoro-5-(5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl)aniline
Kvantitet 500 mg
InChI formel InChI=1S/C13H14F2N4/c14-9-7-10(15)11(16)6-8(9)13-18-17-12-4-2-1-3-5-19(12)13/h6-7H,1-5,16H2
Formel vikt 264.1187