missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-5-methylbenzonitrile, 97%, Thermo Scientific™

Catalog Number p-7052408
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
329314-68-3
C8H5F2N
153.132
MFCD09835341
QUUFHRSTBTZWMA-UHFFFAOYSA-N
2,4-difluoro-5-methyl-benzonitrile, benzonitrile, 2,4-difluoro-5-methyl, pubchem8339, 2,4-difluoro-5-methyl benzonitrile, 2,4-difluoro-5-methylbenzoni trile, benzonitrile, 2,4-difluoro-5-methyl-9ci
10749381
2,4-difluoro-5-methylbenzonitrile
CC1=C(C=C(C(=C1)C#N)F)F
This item is not returnable. View return policy
329314-68-3
C8H5F2N
153.132
MFCD09835341
QUUFHRSTBTZWMA-UHFFFAOYSA-N
2,4-difluoro-5-methyl-benzonitrile, benzonitrile, 2,4-difluoro-5-methyl, pubchem8339, 2,4-difluoro-5-methyl benzonitrile, 2,4-difluoro-5-methylbenzoni trile, benzonitrile, 2,4-difluoro-5-methyl-9ci
10749381
2,4-difluoro-5-methylbenzonitrile
CC1=C(C=C(C(=C1)C#N)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 43°C to 50°C
Color Cream to Yellow
Quantity 250 mg
UN Number UN3439
Formula Weight 153.13
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material 2,4-Difluoro-5-methylbenzonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'