missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoroaniline, 99%, Thermo Scientific Chemicals

Catalog Number p-7021367
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
367-25-9
C6H5F2N
129.11
MFCD00007648
CEPCPXLLFXPZGW-UHFFFAOYSA-N
2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine
9709
2,4-difluoroaniline
NC1=CC=C(F)C=C1F
This item is not returnable. View return policy
367-25-9
C6H5F2N
129.11
MFCD00007648
CEPCPXLLFXPZGW-UHFFFAOYSA-N
2,4-difluorobenzenamine, benzenamine, 2,4-difluoro, 2,4-difluroaniline, 2,4-difluorophenylamine, aniline, 2,4-difluoro, unii-40p93l7kwd, 1-amino-2,4-difluorobenzene, ccris 4621, 2,4-difluoro aniline, 2,4-difluoro-phenylamine
9709
2,4-difluoroaniline
NC1=CC=C(F)C=C1F

2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Difluoroaniline is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -8°C to -7°C
Density 1.281
Boiling Point 168°C to 170°C
Flash Point 62°C (143°F)
Odor Characteristic
Refractive Index 1.507
Quantity 25 g
UN Number UN2941
Beilstein 2802556
Merck Index 14,3147
Solubility Information Not miscible or difficult to mix.
Formula Weight 129.11
Percent Purity 99%
Chemical Name or Material 2,4-Difluoroaniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'