Läs mer
2,4-Difluorobenzoyl Risperidone Impurity, TRC
Produktkod. 17924545
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17924545 25 mg
17961925 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17924545 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44571525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 158697-67-7
Molekylformel C23 H27 F2 N3 O2
Molekylvikt (g/mol) 415.48
Synonym 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(2,4-difluorobenzoyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-, 3-[2-[4-(2,4-Difluorobenzoyl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(2,4-Difluorobenzoyl)piperidino]ethyl]-6,7,8,9-tetrahydro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(2,4-Difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one, Risperidone Imp. H (EP), Risperidone Difluoroketone
IUPAC-namn 3-[2-[4-(2,4-difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
LEDER CC1=C(CCN2CCC(CC2)C(=O)c3ccc(F)cc3F)C(=O)N4CCCCC4=N1
Specifikationer
Kemiskt namn eller material 3-[2-[4-(2,4-Difluorobenzoyl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
Kvantitet 25 mg
InChI formel InChI=1S/C23H27F2N3O2/c1-15-18(23(30)28-10-3-2-4-21(28)26-15)9-13-27-11-7-16(8-12-27)22(29)19-6-5-17(24)14-20(19)25/h5-6,14,16H,2-4,7-13H2,1H3
Formel vikt 415.2071