Läs mer
2',4'-Difluorobiphenyl-4-ol, TRC
Produktkod. 30373285
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30373285 100 mg
30377687 1 g
30385773 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30373285 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD447370100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 59089-68-8
Molekylformel C12H8F2O
Molekylvikt (g/mol) 206.19
Synonym 2',4'-Difluoro[1,1'-biphenyl]-4-ol, 2,4-Difluoro-4'-hydroxybiphenyl, 2',4'-Difluoro-4-hydroxybiphenyl, 4-(2,4-Difluorophenyl)phenol, Diflunisal EP Impurity B
LEDER Oc1ccc(cc1)c2ccc(F)cc2F
Specifikationer
Kemiskt namn eller material 2',4'-Difluorobiphenyl-4-ol
Kvantitet 100 mg
InChI formel InChI=1S/C12H8F2O/c13-9-3-6-11(12(14)7-9)8-1-4-10(15)5-2-8/h1-7,15H
Formel vikt 206.0543