missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dihydroxyacetophenone, 98%, Thermo Scientific Chemicals

Catalog Number p-7081889
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
89-84-9
C8H8O3
152.15
MFCD00002279
SULYEHHGGXARJS-UHFFFAOYSA-N
2',4'-dihydroxyacetophenone, 2,4-dihydroxyacetophenone, 1-2,4-dihydroxyphenyl ethanone, resacetophenone, 4-acetylresorcinol, resoacetophenone, ethanone, 1-2,4-dihydroxyphenyl, 1-acetyl-2,4-dihydroxybenzene, resorcinol, 4-acetyl, 1-2,4-dihydroxyphenyl ethan-1-one
6990
CHEBI:18414
1-(2,4-dihydroxyphenyl)ethanone
CC(=O)C1=CC=C(O)C=C1O
This item is not returnable. View return policy
89-84-9
C8H8O3
152.15
MFCD00002279
SULYEHHGGXARJS-UHFFFAOYSA-N
2',4'-dihydroxyacetophenone, 2,4-dihydroxyacetophenone, 1-2,4-dihydroxyphenyl ethanone, resacetophenone, 4-acetylresorcinol, resoacetophenone, ethanone, 1-2,4-dihydroxyphenyl, 1-acetyl-2,4-dihydroxybenzene, resorcinol, 4-acetyl, 1-2,4-dihydroxyphenyl ethan-1-one
6990
CHEBI:18414
1-(2,4-dihydroxyphenyl)ethanone
CC(=O)C1=CC=C(O)C=C1O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 142°C to 146°C
Quantity 50 g
Beilstein 1282505
Merck Index 14,8140
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 2',4'-Dihydroxyacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'