missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxyquinoline, 97%, Thermo Scientific Chemicals

Catalog Number p-7025323
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
86-95-3
C9H7NO2
161.16
MFCD00006744
HDHQZCHIXUUSMK-UHFFFAOYSA-N
2,4-quinolinediol, 2,4-dihydroxyquinoline, 4-hydroxyquinolin-2 1h-one, quinoline-2,4-diol, 4-hydroxy-2-quinolone, 4-hydroxycarbostyril, 2 1h-quinolinone, 4-hydroxy, 4-hydroxy-2-quinolinone, hydroxycarbostyril, 2-hydroxyquinolin-4 1h-one
54680871
CHEBI:75926
4-hydroxy-1H-quinolin-2-one
C1=CC=C2C(=C1)C(=CC(=O)N2)O
This item is not returnable. View return policy
86-95-3
C9H7NO2
161.16
MFCD00006744
HDHQZCHIXUUSMK-UHFFFAOYSA-N
2,4-quinolinediol, 2,4-dihydroxyquinoline, 4-hydroxyquinolin-2 1h-one, quinoline-2,4-diol, 4-hydroxy-2-quinolone, 4-hydroxycarbostyril, 2 1h-quinolinone, 4-hydroxy, 4-hydroxy-2-quinolinone, hydroxycarbostyril, 2-hydroxyquinolin-4 1h-one
54680871
CHEBI:75926
4-hydroxy-1H-quinolin-2-one
C1=CC=C2C(=C1)C(=CC(=O)N2)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 5 g
Beilstein 129767
Solubility Information Insoluble in water
Formula Weight 161.16
Percent Purity 97%
Chemical Name or Material 2,4-Dihydroxyquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'