missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxyphenylacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 108°C to 112°C |
Quantity | 1 g |
Beilstein | 2694132 |
Synonym | 2,4-dimethoxyphenylacetic acid, 2-2,4-dimethoxyphenyl acetic acid, 2,4-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,4-dimethoxy, 2,4-dimethoxybenzeneacetic acid, acmc-209nop, ksc497q2d, 2,4-dimethoxyphenylaceticacid, 2, 4-dimethoxyphenylacetic acid |
Formula Weight | 196.2 |
Percent Purity | 98% |
Chemical Name or Material | 2,4-Dimethoxyphenylacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'