Läs mer
2,4-Dimethyl-1-heptene, TRC
Produktkod. 30376115
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30376115 50 mg
30376221 500 mg
30383573 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30376115 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD46029850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19549-87-2
Molekylformel C9H18
Molekylvikt (g/mol) 126.23
Synonym 2,4-Dimethyl-1-heptene
LEDER CCCC(C)CC(=C)C
Specifikationer
Kemiskt namn eller material 2,4-Dimethyl-1-heptene
Kvantitet 50 mg
InChI formel InChI=1S/C9H18/c1-5-6-9(4)7-8(2)3/h9H,2,5-7H2,1,3-4H3
Formel vikt 126.1409