Läs mer
(2,4-Dioxo-thiazolidin-3-yl)acetic Acid, TRC
Produktkod. 30179966
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179966 100 mg
30179919 50 mg
30180177 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179966 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD487080100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 31061-24-2
Molekylformel C5H5NO4S
Molekylvikt (g/mol) 175.16
Synonym (2,4-Dioxo-thiazolidin-3-yl)-acetic acid
LEDER OC(=O)CN1C(=O)CSC1=O
Specifikationer
Kemiskt namn eller material (2,4-Dioxo-thiazolidin-3-yl)acetic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C5H5NO4S/c7-3-2-11-5(10)6(3)1-4(8)9/h1-2H2,(H,8,9)
Formel vikt 174.9939