missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Fluorophenyl)-5-nitrobenzimidazole, 95%

Brand:  Alfa Aesar™ H64682.06

Additional Details : CAS Number : 348-35-6 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15451989

  • 13699.94 SEK / 5g


Estimated Shipment date
from Supplier 23-01-2019
Add to basket

Specifications

Chemical Identifiers

348-35-6
257.224
FAHRDVODAHLFLE-UHFFFAOYSA-N
4915462
C1=CC(=CC=C1C2=NC3=C(N2)C=C(C=C3)[N+](=O)[O-])F
C13H8FN3O2
MFCD13812378
2-4-fluorophenyl-6-nitro-1h-benzimidazole, 2-4-fluoro-phenyl-5-nitro-1h-benzimidazole, 2-4-fluorophenyl-5-nitro-3h-1,3-benzodiazole, 2-4-fluorophenyl-5-nitrobenzimidazole, 2-4-fluorophenyl-5-nitro-1h-benzoimidazole, 2-4-fluorophenyl-6-nitro-1h-benzo d imidazole, 2-4-fluorophenyl-5-nitro-1h-1,3-benzodiazole
2-(4-fluorophenyl)-6-nitro-1H-benzimidazole

Specifications

2-(4-Fluorophenyl)-5-nitrobenzimidazole
348-35-6
MFCD13812378
FAHRDVODAHLFLE-UHFFFAOYSA-N
2-(4-fluorophenyl)-6-nitro-1H-benzimidazole
4915462
Solid
5g
C13H8FN3O2
2-4-fluorophenyl-6-nitro-1h-benzimidazole, 2-4-fluoro-phenyl-5-nitro-1h-benzimidazole, 2-4-fluorophenyl-5-nitro-3h-1,3-benzodiazole, 2-4-fluorophenyl-5-nitrobenzimidazole, 2-4-fluorophenyl-5-nitro-1h-benzoimidazole, 2-4-fluorophenyl-6-nitro-1h-benzo d imidazole, 2-4-fluorophenyl-5-nitro-1h-1,3-benzodiazole
C1=CC(=CC=C1C2=NC3=C(N2)C=C(C=C3)[N+](=O)[O-])F
257.224
257.22
95%
Videos
SDS