Läs mer
2-(4-Hydroxyphenyl)-3-methyl-1H-indol-5-ol, TRC
Produktkod. 17953345
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17953345 10 mg
17921835 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17953345 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95140010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 91444-54-1
Molekylformel C15 H13 N O2
Molekylvikt (g/mol) 239.27
Synonym 2-(4-Hydroxy-Phenyl)-3-Methyl-1H-Indole-5-Ol
IUPAC-namn 2-(4-hydroxyphenyl)-3-methyl-1H-indol-5-ol
LEDER Cc1c([nH]c2ccc(O)cc12)c3ccc(O)cc3
Specifikationer
Kemiskt namn eller material 2-(4-Hydroxyphenyl)-3-methyl-1H-indol-5-ol
Kvantitet 10 mg
InChI formel InChI=1S/C15H13NO2/c1-9-13-8-12(18)6-7-14(13)16-15(9)10-2-4-11(17)5-3-10/h2-8,16-18H,1H3
Formel vikt 239.0946