Läs mer
2-(4-Hydroxyphenyl)-N,N-dimethylethylamine Hydrochloride, TRC
Produktkod. 30404079
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30404079 500 mg
1 options
Produktkod. 30404079 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH973108500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hordenine Hydrochloride (4-Hydroxy-N,N-dimethylphenethylamine Hydrochloride)
CAS 6027-23-2
Molekylformel C10 H15 N O . Cl H
Kvantitet 500 mg
Synonym Hordenine Hydrochloride, 4-Hydroxy-N,N-dimethylphenethylamine Hydrochloride, Phenol, 4-[2-(dimethylamino)ethyl]-, hydrochloride (1:1), Hordenine hydrochloride (6CI), Phenol, 4-[2-(dimethylamino)ethyl]-, hydrochloride (9CI), Phenol, p-[2-(dimethylamino)ethyl]-, hydrochloride (8CI), N,N-Dimethyltyramine hydrochloride
InChI formel InChI=1S/C10H15NO.ClH/c1-11(2)8-7-9-3-5-10(12)6-4-9;/h3-6,12H,7-8H2,1-2H3;1H
LEDER Cl.CN(C)CCc1ccc(O)cc1
IUPAC-namn 4-[2-(dimethylamino)ethyl]phenol;hydrochloride
Molekylvikt (g/mol) 201.69
Formel vikt 201.092