Läs mer
2-(4-Methoxybenzoyl)-N,N-bis(3-methylbutyl)-1-(3-piperidin-1-ylpropyl)-1H-benzimidazole-6-carboxamide hydrochloride, TRC
Produktkod. 30468487
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30468487 100 mg
1 options
Produktkod. 30468487 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM363680100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(4-methoxybenzoyl)-N,N-bis(3-methylbutyl)-1-(3-piperidin-1-ylpropyl)-1H-benzimidazole-6-carboxamide hydrochloride
CAS 852803-39-5
Molekylformel C34 H48 N4 O3 . xHCl
Kvantitet 100 mg
Synonym 1H-Benzimidazole-6-carboxamide, 2-(4-methoxybenzoyl)-N,N-bis(3-methylbutyl)-1-[3-(1-piperidinyl)propyl]-, hydrochloride (1:?) (ACI), 1H-Benzimidazole-6-carboxamide, 2-(4-methoxybenzoyl)-N,N-bis(3-methylbutyl)-1-[3-(1-piperidinyl)propyl]-, hydrochloride (9CI)
InChI formel InChI=1S/C34H48N4O3.ClH/c1-25(2)16-22-37(23-17-26(3)4)34(40)28-12-15-30-31(24-28)38(21-9-20-36-18-7-6-8-19-36)33(35-30)32(39)27-10-13-29(41-5)14-11-27;/h10-15,24-26H,6-9,16-23H2,1-5H3;1H
LEDER O=C(N(CCC(C)C)CCC(C)C)C1=CC=C2C(N(C(C(C3=CC=C(C=C3)OC)=O)=N2)CCCN4CCCCC4)=C1.Cl
IUPAC-namn N,N-diisopentyl-2-(4-methoxybenzoyl)-3-[3-(1-piperidyl)propyl]benzimidazole-5-carboxamide;hydrochloride
Molekylvikt (g/mol) 597.2