Läs mer
2-(4-Methoxyphenyl)ethylamine, TRC
Produktkod. 30477404
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30477404 1 g
1 options
Produktkod. 30477404 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2660751G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(4-Methoxyphenyl)ethylamine
CAS 55-81-2
Molekylformel C9 H13 N O
Kvantitet 1 g
Synonym Benzeneethanamine, 4-methoxy-, Phenethylamine, p-methoxy- (6CI,8CI), 4-Methoxybenzeneethanamine, 1-(4-Methoxybenzyl)-N-methylamine, 2-(4-Methoxyphenyl)ethan-1-amine, 2-(4-Methoxyphenyl)ethanamine, 2-(4-Methoxyphenyl)ethylamine, 2-(p-Methoxyphenyl)ethylamine, 4-Methoxy-β-phenethylamine, 4-Methoxy-β-phenylethylamine, 4-Methoxyphenethylamine, 4-Methoxyphenylethylamine, Homoanisylamine, NSC 43687, O-Methyltyramine, Tyramine methyl ether, Tyramine, O-methyl-, [2-[4-(Methyloxy)phenyl]ethyl]amine, p-Methoxyphenethylamine
InChI formel InChI=1S/C9H13NO/c1-11-9-4-2-8(3-5-9)6-7-10/h2-5H,6-7,10H2,1H3
LEDER COc1ccc(CCN)cc1
IUPAC-namn 2-(4-methoxyphenyl)ethanamine
Molekylvikt (g/mol) 151.21
Formel vikt 151.0997