missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Methoxyphenyl)-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide, 95%

Brand:  Alfa Aesar™ H65863.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15449956

  • 2362.17 SEK / 1g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

2-(4-Methoxyphenyl)-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide
295.38
MFCD01418412

Specifications

2-(4-Methoxyphenyl)-N-(5,6,7,8-tetrahydro-1-naphthyl)acetamide
95%
MFCD01418412
Solid
295.38
1g
Videos
SDS