Läs mer
2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride, Mikromol™
Produktkod. 30051599
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30051599 25 mg
30051208 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30051599 Leverantör Mikromol Leverantörsnummer MM0393.020025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride
CAS 50822-98-5
Analyt- eller komponentnamn 2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride
Frakt skick Room Temperature
Molekylformel C11 H17 N O . Cl H
Certifikat och efterlevnad ISO 17025
Synonym Venlafaxine Hydrochloride Imp. A (EP), Venlafaxine Imp. A (EP), 2-(4-Methoxyphenyl)-N,N-dimethylethanamine Hydrochloride, Venlafaxine Hydrochloride Imp. A (EP) as Hydrochloride, Venlafaxine Hydrochloride Impurity A as Hydrochloride, Venlafaxine Impurity A as Hydrochloride
InChI formel InChI=1S/C11H17NO.ClH/c1-12(2)9-8-10-4-6-11(13-3)7-5-10;/h4-7H,8-9H2,1-3H3;1H
LEDER Cl.COc1ccc(CCN(C)C)cc1
IUPAC-namn 2-(4-methoxyphenyl)-N,N-dimethylethanamine;hydrochloride
Molekylvikt (g/mol) 215.72
Kvantitet 25 mg
Formel vikt 215.1077 g/mol