missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(4-Methoxyphenyl)thiazolidine-4-carboxylic acid, 97%

Brand:  Alfa Aesar™ H62065.03

Additional Details : CAS Number : 65884-40-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15496976

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

65884-40-4
239.289
ZQRSXNJVEHLFCE-UHFFFAOYSA-N
2827728
COC1=CC=C(C=C1)C2NC(CS2)C(=O)O
C11H13NO3S
MFCD00627014
2-4-methoxyphenyl-1,3-thiazolidine-4-carboxylic acid, 2-4-methoxy-phenyl-thiazolidine-4-carboxylic acid, 2-4-methoxyphenyl thiazolidine-4-carboxylic acid, 2-4-methoxyphenyl-4-carboxy-1,3-thiazolidine, 2-4-methoxy-phenyl thiazolidine-4-carboxylic acid, 2-4-methoxyphenyl-thiazolidine-4-carboxylic acid, 2-p-methoxyphenyl-1,3-thiazolidine-4-carboxylic acid
2-(4-methoxyphenyl)-1,3-thiazolidine-4-carboxylic acid

Specifications

2-(4-Methoxyphenyl)thiazolidine-4-carboxylic acid
65884-40-4
MFCD00627014
ZQRSXNJVEHLFCE-UHFFFAOYSA-N
2-(4-methoxyphenyl)-1,3-thiazolidine-4-carboxylic acid
2827728
Solid
1g
C11H13NO3S
2-4-methoxyphenyl-1,3-thiazolidine-4-carboxylic acid, 2-4-methoxy-phenyl-thiazolidine-4-carboxylic acid, 2-4-methoxyphenyl thiazolidine-4-carboxylic acid, 2-4-methoxyphenyl-4-carboxy-1,3-thiazolidine, 2-4-methoxy-phenyl thiazolidine-4-carboxylic acid, 2-4-methoxyphenyl-thiazolidine-4-carboxylic acid, 2-p-methoxyphenyl-1,3-thiazolidine-4-carboxylic acid
COC1=CC=C(C=C1)C2NC(CS2)C(=O)O
239.289
239.29
97%
Videos
SDS Documents