Learn More
Alfa Aesar™ 2-(4-Methoxyphenyl)thiazolidine, 97%
Brand:  Alfa Aesar™ H62999.03
Additional Details : CAS Number : 31404-08-7 Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15499418
  • 1007.07 SEK / 1g
Estimated Shipment date
from Supplier 20-02-2019
Add to basket
Specifications
Chemical Identifiers
31404-08-7
195.28
OQFUUZFCTONCOU-UHFFFAOYSA-N
98272
COC1=CC=C(C=C1)C2NCCS2
C10H13NOS
MFCD00222821
2-4-methoxyphenyl-1,3-thiazolane, 2-4-methoxy-phenyl-thiazolidine, 2-4-methoxyphenyl thiazolidine, 2-4-methoxyphenyl-1,3-thiazolidine, thiazolidine, 2-4-methoxyphenyl, 4-methoxy-1-1,3-thiazolidin-2-yl benzene, methoxyphenylthiazolane, 4-27-00-01814 beilstein handbook reference, 2-4-methoxyphenyl thiazolidine
2-(4-methoxyphenyl)-1,3-thiazolidine
Specifications
2-(4-Methoxyphenyl)thiazolidine
31404-08-7
MFCD00222821
OQFUUZFCTONCOU-UHFFFAOYSA-N
2-(4-methoxyphenyl)-1,3-thiazolidine
98272
Solid
1g
C10H13NOS
2-4-methoxyphenyl-1,3-thiazolane, 2-4-methoxy-phenyl-thiazolidine, 2-4-methoxyphenyl thiazolidine, 2-4-methoxyphenyl-1,3-thiazolidine, thiazolidine, 2-4-methoxyphenyl, 4-methoxy-1-1,3-thiazolidin-2-yl benzene, methoxyphenylthiazolane, 4-27-00-01814 beilstein handbook reference, 2-4-methoxyphenyl thiazolidine
COC1=CC=C(C=C1)C2NCCS2
195.28
195.28
97%