Läs mer
2-(4-methylphenyl)oxirane, TRC
Produktkod. 30464962
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30464962 10 mg
30471232 50 mg
30473109 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30464962 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM35548810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13107-39-6
Molekylformel C9H10O
Molekylvikt (g/mol) 134.17
IUPAC-namn 2-(4-methylphenyl)oxirane
LEDER Cc1ccc(cc1)C2CO2
Specifikationer
Kemiskt namn eller material 2-(4-methylphenyl)oxirane
Kvantitet 10 mg
InChI formel InChI=1S/C9H10O/c1-7-2-4-8(5-3-7)9-6-10-9/h2-5,9H,6H2,1H3
Formel vikt 134.0732