Learn More
2-(4-Methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde, 97%, Thermo Scientific™
Catalog Number p-3629687
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
320423-50-5
C9H13N3OS
211.283
MFCD01568852
TUJAFVJUJXMFEG-UHFFFAOYSA-N
2-4-methylpiperazin-1-yl-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazino-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carbaldehyde, 2-4-methyl-1-piperazinyl thiazole-5-carboxaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carboxaldehyde, 2-4-methylpiperazinyl-1,3-thiazole-5-carbaldehyde, acmc-20aoov, methylpiperazinothiazolecarbaldehyde
1490234
2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde
CN1CCN(CC1)C2=NC=C(S2)C=O
320423-50-5
C9H13N3OS
211.283
MFCD01568852
TUJAFVJUJXMFEG-UHFFFAOYSA-N
2-4-methylpiperazin-1-yl-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazino-1,3-thiazole-5-carbaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carbaldehyde, 2-4-methyl-1-piperazinyl thiazole-5-carboxaldehyde, 2-4-methylpiperazin-1-yl thiazole-5-carboxaldehyde, 2-4-methylpiperazinyl-1,3-thiazole-5-carbaldehyde, acmc-20aoov, methylpiperazinothiazolecarbaldehyde
1490234
2-(4-methylpiperazin-1-yl)-1,3-thiazole-5-carbaldehyde
CN1CCN(CC1)C2=NC=C(S2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 211.29
Percent Purity 97%
Chemical Name or Material 2-(4-Methylpiperazin-1-yl)-1, 3-thiazole-5-carbaldehyde