missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylpiperazino)benzaldehyde, ≥95%, Thermo Scientific™

Catalog Number p-3110165
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
85803-62-9
C12H16N2O
204.273
MFCD01313808
GSRYZPWIWYYROI-UHFFFAOYSA-N
2-4-methylpiperazin-1-yl benzaldehyde, 2-4-methylpiperazino benzaldehyde, benzaldehyde,2-4-methyl-1-piperazinyl, 4-methylpiperazin-1-ylbenzaldehyde, 2-4-methylpiperazinyl benzaldehyde, 2-4-methylpiperazinyl-benzaldehyde, 2-4-methyl-1-piperazinyl benzaldehyde, 2-4methyl-1-piperazinyl-benzaldehyde, 2-4-methyl-1-piperazinyl-benzaldehyde, 2-4-methylpiperazin-1-yl-benzaldehyde
2795566
2-(4-methylpiperazin-1-yl)benzaldehyde
CN1CCN(CC1)C2=CC=CC=C2C=O
This item is not returnable. View return policy
85803-62-9
C12H16N2O
204.273
MFCD01313808
GSRYZPWIWYYROI-UHFFFAOYSA-N
2-4-methylpiperazin-1-yl benzaldehyde, 2-4-methylpiperazino benzaldehyde, benzaldehyde,2-4-methyl-1-piperazinyl, 4-methylpiperazin-1-ylbenzaldehyde, 2-4-methylpiperazinyl benzaldehyde, 2-4-methylpiperazinyl-benzaldehyde, 2-4-methyl-1-piperazinyl benzaldehyde, 2-4methyl-1-piperazinyl-benzaldehyde, 2-4-methyl-1-piperazinyl-benzaldehyde, 2-4-methylpiperazin-1-yl-benzaldehyde
2795566
2-(4-methylpiperazin-1-yl)benzaldehyde
CN1CCN(CC1)C2=CC=CC=C2C=O
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 204.27
Percent Purity 95%
Chemical Name or Material 2-(4-Methylpiperazin-1-yl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'