Läs mer
2-(4-Methylthiazol-5-yl)ethanamine, TRC
Produktkod. 30474413
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474413 100 mg
30473559 50 mg
30480451 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30474413 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM227715100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 58981-35-4
Molekylformel C6H10N2S
Molekylvikt (g/mol) 142.22
Synonym 2-(4-Methyl-1,3-thiazol-5-yl)ethan-1-amine, 2-(4-Methyl-1,3-thiazol-5-yl)ethanamine, VUF 8952
IUPAC-namn 2-(4-methyl-1,3-thiazol-5-yl)ethanamine
LEDER Cc1ncsc1CCN
Specifikationer
Kemiskt namn eller material 2-(4-Methylthiazol-5-yl)ethanamine
Kvantitet 100 mg
InChI formel InChI=1S/C6H10N2S/c1-5-6(2-3-7)9-4-8-5/h4H,2-3,7H2,1H3
Formel vikt 142.0565