Läs mer
2-(4-Nitro-1-oxoisoindolin-2-yl)pentanedioic Acid, TRC
Produktkod. 30469004
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30469004 50 mg
30471463 100 mg
30466423 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30469004 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN49690350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 295357-72-1
Molekylformel C13H12N2O7
Molekylvikt (g/mol) 308.25
Synonym 2-(1,3-Dihydro-4-nitro-1-oxo-2H-isoindol-2-yl)pentanedioic Acid
IUPAC-namn 2-(7-nitro-3-oxo-1H-isoindol-2-yl)pentanedioic acid
LEDER OC(=O)CCC(N1Cc2c(cccc2[N+](=O)[O-])C1=O)C(=O)O
Specifikationer
Kemiskt namn eller material 2-(4-Nitro-1-oxoisoindolin-2-yl)pentanedioic Acid
Kvantitet 50 mg
InChI formel InChI=1S/C13H12N2O7/c16-11(17)5-4-10(13(19)20)14-6-8-7(12(14)18)2-1-3-9(8)15(21)22/h1-3,10H,4-6H2,(H,16,17)(H,19,20)
Formel vikt 308.0645