Läs mer
2-(4-Octylphenyl)ethyl Acetate, TRC
Produktkod. 30414875
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30414875 50 mg
1 options
Produktkod. 30414875 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO29383050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(4-Octylphenyl)ethyl Acetate
CAS 162358-04-5
Molekylformel C18 H28 O2
Kvantitet 50 mg
Synonym Benzeneethanol','4-octyl-','1-acetate, Benzeneethanol','4-octyl-','acetate (9CI), 4-Octylphenethyl acetate, 2-(4-Octylphenyl)ethyl acetate
InChI formel InChI=1S/C18H28O2/c1-3-4-5-6-7-8-9-17-10-12-18(13-11-17)14-15-20-16(2)19/h10-13H,3-9,14-15H2,1-2H3
LEDER CCCCCCCCc1ccc(CCOC(=O)C)cc1
IUPAC-namn 2-(4-octylphenyl)ethyl acetate
Molekylvikt (g/mol) 276.41
Formel vikt 276.2089