Läs mer
2-(4-tert-Butylphenyl)acetonitrile, TRC
Produktkod. 30342863
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30342863 5 g
1 options
Produktkod. 30342863 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6944355G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-(4-tert-Butylphenyl)acetonitrile
CAS 3288-99-1
Molekylformel C12H15N
Kvantitet 5 g
Synonym (4-tert-Butylphenyl)acetonitrile, 4-(1,1-Dimethylethyl)benzeneacetonitrile (ACI), Acetonitrile, (p-tert-butylphenyl)- (6CI, 7CI, 8CI), (4-tert-Butylphenyl)acetonitrile, (p-tert-Butylphenyl)acetonitrile, 2-(4-tert-Butylphenyl)acetonitrile, 2-[4-(tert-Butyl)phenyl]ethanenitrile, 4-tert-Butylbenzyl cyanide, NSC 85354, p-tert-Butylbenzyl cyanide, Benzeneacetonitrile, 4-(1,1-dimethylethyl)- (9CI, ACI)
InChI formel InChI=1S/C12H15N/c1-12(2,3)11-6-4-10(5-7-11)8-9-13/h4-7H,8H2,1-3H3
LEDER CC(C)(C)c1ccc(CC#N)cc1
IUPAC-namn 2-(4-tert-butylphenyl)acetonitrile
Molekylvikt (g/mol) 173.25
Formel vikt 173.1204