Läs mer
2,4-Xylenesulfonic Acid Hydrate, TRC
Produktkod. 30452245
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30452245 5 g
1 options
Produktkod. 30452245 Leverantör Toronto Research Chemicals Leverantörsnummer TRCX7494955G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2,4-Xylenesulfonic Acid Hydrate
CAS 58811-92-0
Molekylformel C8 H10 O3 S . H2 O
Kvantitet 5 g
Synonym 2,4-Dimethylbenzenesulfonic Acid Hydrate, SXA 60 Hydrate, m-Xylene-4-sulfonic Acid Hydrate, m-Xylenesulfonic Acid Hydrate
InChI formel InChI=1S/C8H10O3S.H2O/c1-6-3-4-8(7(2)5-6)12(9,10)11;/h3-5H,1-2H3,(H,9,10,11);1H2
LEDER O.Cc1ccc(c(C)c1)S(=O)(=O)O
IUPAC-namn 2,4-dimethylbenzenesulfonic acid;hydrate
Molekylvikt (g/mol) 204.243
Formel vikt 204.0456