Läs mer
2-((5(6)-Tetramethyl-rhodamine)carboxylamino)ethyl Methanethiosulfonate (90%), TRC
Produktkod. 30452968
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452968 10 mg
30454010 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30452968 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT30517510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1329837-19-5
Molekylformel C28H29N3O6S2
Molekylvikt (g/mol) 567.68
Synonym MTS-TAMRA
LEDER CC(=O)NCCSS(=O)(=O)C.CN(C)C1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)N(C)C)C5=CC=CC=C5C(=O)O3
Specifikationer
Kemiskt namn eller material 2-((5(6)-Tetramethyl-rhodamine)carboxylamino)ethyl Methanethiosulfonate
Kvantitet 10 mg
InChI formel InChI=1S/C24H22N2O3.C5H11NO3S2/c1-25(2)15-9-11-19-21(13-15)28-22-14-16(26(3)4)10-12-20(22)24(19)18-8-6-5-7-17(18)23(27)29-24;1-5(7)6-3-4-10-11(2,8)9/h5-14H,1-4H3;3-4H2,1-2H3,(H,6,7)