missing translation for 'onlineSavingsMsg'
Läs mer

2-((5(6)-Tetramethyl-rhodamine)carboxylamino)ethyl Methanethiosulfonate (90%), TRC

Produktkod. 30454010
Change view
Click to view available options
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30454010 1 mg
30452968 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30454010 Supplier Toronto Research Chemicals Supplier No. TRCT3051751MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1329837-19-5
Molekylformel C28H29N3O6S2
Molekylvikt (g/mol) 567.68
Synonym MTS-TAMRA
LEDER CC(=O)NCCSS(=O)(=O)C.CN(C)C1=CC2=C(C=C1)C3(C4=C(O2)C=C(C=C4)N(C)C)C5=CC=CC=C5C(=O)O3

Specifications

Kemiskt namn eller material 2-((5(6)-Tetramethyl-rhodamine)carboxylamino)ethyl Methanethiosulfonate
Kvantitet 1 mg
InChI formel InChI=1S/C24H22N2O3.C5H11NO3S2/c1-25(2)15-9-11-19-21(13-15)28-22-14-16(26(3)4)10-12-20(22)24(19)18-8-6-5-7-17(18)23(27)29-24;1-5(7)6-3-4-10-11(2,8)9/h5-14H,1-4H3;3-4H2,1-2H3,(H,6,7)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.