Läs mer
[2-[5-(Aminomethyl)-2-methyl-1H-imidazol-1-yl]-5-chlorophenyl](2-fluorophenyl)-methanone Dihydrochloride, TRC
Produktkod. 30320854
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30320854 250 mg
1 options
Produktkod. 30320854 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA616235250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (2-[5-(Aminomethyl)-2-methyl-1H-imidazol-1-yl]-5-chlorophenyl)(2-fluorophenyl)-methanone Dihydrochloride
CAS 59468-73-4
Molekylformel C18H15ClFN3O • 2(HCl)
Kvantitet 250 mg
Synonym Methanone, [2-[5-(aminomethyl)-2-methyl-1H-imidazol-1-yl]-5-chlorophenyl](2-fluorophenyl)-, dihydrochloride (9CI)
InChI formel InChI=1S/C18H15ClFN3O.2ClH/c1-11-22-10-13(9-21)23(11)17-7-6-12(19)8-15(17)18(24)14-4-2-3-5-16(14)20;;/h2-8,10H,9,21H2,1H3;2*1H
LEDER Cl.Cl.Cc1ncc(CN)n1c2ccc(Cl)cc2C(=O)c3ccccc3F
IUPAC-namn [2-[5-(aminomethyl)-2-methylimidazol-1-yl]-5-chlorophenyl]-(2-fluorophenyl)methanone;dihydrochloride
Molekylvikt (g/mol) 343.78
Formel vikt 415.0421