Läs mer
2,5-Anhydro-3,4-dibenzyl-D-glucitol, TRC
Product Code. 30335807
Change view
Click to view available options
Kvantitet:
25 mg
Product Code. Kvantitet
30335807 25 mg
1 options
Product Code. 30335807 Supplier Toronto Research Chemicals Supplier No. TRCA63785025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 2,5-Anhydro-3,4-dibenzyl-D-glucitol
CAS 129115-89-5
Molekylformel C20H24O5
Kvantitet 25 mg
Synonym 2,5-Anhydro-3,4-bis-O-(phenylmethyl)-D-glucitol
InChI formel InChI=1S/C20H24O5/c21-11-17-19(23-13-15-7-3-1-4-8-15)20(18(12-22)25-17)24-14-16-9-5-2-6-10-16/h1-10,17-22H,11-14H2/t17-,18+,19-,20-/m1/s1
LEDER OC[C@@H]1O[C@H](CO)[C@@H](OCc2ccccc2)[C@@H]1OCc3ccccc3
IUPAC-namn [(2R,3R,4R,5S)-5-(hydroxymethyl)-3,4-bis(phenylmethoxy)oxolan-2-yl]methanol
Molekylvikt (g/mol) 344.4
Formel vikt 344.1624