Läs mer
2,5-Anhydro-D-glucitol-1,6-diphosphate, TRC
Produktkod. 30012476
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012476 5 mg
30003379 10 mg
30004136 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012476 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA6480205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4429-47-4
Molekylformel C6 H14 O11 P2
Molekylvikt (g/mol) 324.12
IUPAC-namn [(2S,3S,4S,5R)-3,4-dihydroxy-5-(phosphonooxymethyl)oxolan-2-yl]methyl dihydrogen phosphate
LEDER O[C@H]1[C@H](O)[C@@H](COP(=O)(O)O)O[C@H]1COP(=O)(O)O
Specifikationer
Kemiskt namn eller material 2,5-Anhydro-D-glucitol-1,6-diphosphate
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C6H14O11P2/c7-5-3(1-15-18(9,10)11)17-4(6(5)8)2-16-19(12,13)14/h3-8H,1-2H2,(H2,9,10,11)(H2,12,13,14)/t3-,4+,5-,6-/m1/s1
Formel vikt 324.0011
Procent renhet >95%