missing translation for 'onlineSavingsMsg'
Läs mer

2,5-Anhydro-D-glucitol, TRC

Produktkod. 30329147
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30329147 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30329147 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA648005100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 2,5-Anhydro-D-glucitol
CAS 27826-73-9
Anteckningar om renhetsgrad HPLC
Molekylformel C6 H12 O5
Kvantitet 100 mg
Synonym D-Glucitol, 2,5-anhydro-, Glucitol, 2,5-anhydro-, D- (8CI), 2,5-Anhydro-D-glucitol, 2,5-Anhydro-D-sorbitol, 2,5-Anhydrosorbitol, 2,5-Sorbitan, (2R,3S,4S,5S)-3,4-Dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran
InChI formel InChI=1S/C6H12O5/c7-1-3-5(9)6(10)4(2-8)11-3/h3-10H,1-2H2/t3-,4+,5-,6-/m1/s1
LEDER OC[C@@H]1O[C@H](CO)[C@@H](O)[C@@H]1O
IUPAC-namn (2R,3S,4S,5S)-2,5-bis(hydroxymethyl)oxolane-3,4-diol
Molekylvikt (g/mol) 164.16
Formel vikt 164.0685
Procent renhet >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.